Understanding Metal Surfaces and Oxides (Recorded Webinar)
This webinar covers the basics of X-ray photoelectron spectroscopy, with a special emphasis on how it can be used in the field of metal and oxide analysis. X-ray photoelectron spectroscopy is a powerful technique for the chemical analysis of the surface of materials.
Requiring little or no sample preparation, and able to directly quantify all elements except hydrogen and helium, XPS delivers chemical state information from the topmost few nanometers of the surface of a sample. Modern instruments, such as the Thermo Scientific K-Alpha, can extend analyses into chemical images of the surface, or depth profiles to reveal the composition of buried interfaces.
These strengths allow scientists and engineers to measure passivation coatings, understand catalyst chemistries, and develop bio-compatibility coatings.
Fill out the form to view the recorded webinar
View the webinar
Fields marked with a * are required.
Email Address *
First Name *
Last Name *
Company *
Street Address *
Country *
State or Province *
City *
Zip/Postal Code *
Business Phone *
How can our team help you?
Would you like a representative to contact you? *
Yes   No
Would you like a product demo? *
Yes   No
Would you like a product quote? *
Yes   No
I would like to receive email alerts about products, events and special offers that match my interest.