Gulf Coast Conference Workshop 2016
Solving Real World Challenges in Spectroscopy and Material Characterization
Join us on for this dynamic, complimentary workshop that takes you from theory to research and industrial applications with FTIR, IR Microscopy, Raman, Rheology and Extrusion technologies. Gain hands-on experience and learn about the latest in instrumentation, software and sample handling techniques.
Save your seat for this FREE workshop at GCC
Please note: the workshop is free but a registration fee is required by GCC for entrance to plenary sessions and workshops.
Agenda | ||
8:00 - 9:00: Material Characterization – Rheology for Polymer and Crude Oil Industries The petrochemical and polymer industries utilize rheology to better understand their materials and how they behave under controlled conditions. This allows users to gain insight into their processes and solve real-world challenges. Relevant applications would include the determination of physical properties (viscoelasticity, molecular weights and distribution, transition temperatures, extensional viscosity), processability (degradation, curing) and flow under extreme conditions (high pressure and temperatures). Here, we will discuss several methods that employ rotational rheometers, extensional rheometers and torque rheometers to address these challenges. | ||
9:15 - 10:15: Infrared Spectroscopy – FTIR Theory, Sample Handling Techniques, Hands-on Training | ||
10:30 - 12:00 Thermo Scientific™ OMNIC Software Tutorial for IR/Raman Spectroscopy This workshop will provide an overview of OMNIC spectroscopy software and how it can be used to collect, manipulate and identify infrared, NIR and Raman spectra. Live samples will be scanned to illustrate key features and capabilities in the software. A question and answer forum can help solve specific challenges that spectroscopists encounter. | ||
1:00 - 1:45 Extrusion Techniques for Polymer and Crude Oil Industries | ||
2:00 - 3:00: Thermo Scientific TQ Analyst™ Software and Macros - Quantitative software training with pre-programmed operation for FTIR A live demonstration of TQ Analyst software will illustrate FTIR, FT-NIR and Raman quantitative and qualitative methods. You will learn how to diagnose chemometric methods to ensure they accurately predict. Some pitfalls will be covered to avoid improperly modeled methods. In addition, an overview of Macros/Basic automated workflow software will be given. This software is designed to build pre-program quantitative predictions. The Macros language can also automate many other software functions, like basic kinetic routines. | ||
3:15 - 4:15: FTIR Spectral Interpretation and Problem Solving | ||
4:15 - 5:00: Micro-Spectroscopic Sampling – FTIR and Raman Microscopy This workshop will highlight the advantages of using Infrared and Raman microscopy techniques in the analysis of micro-scale materials. The presentation will illustrate various applications of both microscopy techniques and the advantages of each based on the type of materials being analyzed. |